Assay Details
| Gene Symbol: per | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272265.1 | NP_001259194.1 | 7 - 8 | 3951 | 66 | |
| NM_080317.2 | NP_525056.2 | 7 - 8 | 3951 | 66 | ||
More Information
Set Membership: |
|

per
Genomic Map