Assay Details
| Gene Symbol: rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169193.2 | NP_001162664.2 | 1 - 2 | 962 | 104 | |
| NM_001272310.1 | NP_001259239.1 | 1 - 2 | 962 | 104 | ||
| NM_080332.4 | NP_525071.2 | 1 - 2 | 962 | 104 | ||
More Information
Set Membership: |
|

rb
Genomic Map