Assay Details
| Gene Symbol: up | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014737.2 | NP_001014737.1 | 4 - 5 | 207 | 67 | |
| NM_001169271.2 | NP_001162742.1 | 4 - 5 | 207 | 67 | ||
| NM_001272594.2 | NP_001259523.1 | 4 - 5 | 204 | 67 | ||
| NM_080349.3 | NP_525088.2 | 4 - 5 | 207 | 67 | ||
| GenBank mRNA | BT125908.1 | - | 4 - 5 | 186 | 67 | |
More Information
Set Membership: |
|

up
Genomic Map