Assay Details
| Gene Symbol: Prosalpha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080353.5 | NP_525092.1 | 2 - 3 | 482 | 71 | |
| GenBank mRNA | AY071241.1 | - | 2 - 3 | 480 | 71 | |
More Information
Set Membership: |
|

Prosalpha4
Genomic Map