Assay Details
| Gene Symbol: bun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042892.3 | NP_001036357.2 | 4 - 5 | 2376 | 114 | |
| NM_001042894.2 | NP_001036359.1 | 4 - 5 | 3143 | 114 | ||
| NM_001298946.1 | NP_001285875.1 | 3 - 4 | 2176 | 114 | ||
| NM_080364.4 | NP_525103.2 | 3 - 4 | 3405 | 114 | ||
More Information
Set Membership: |
|

bun
Genomic Map