Assay Details
| Gene Symbol: bun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042894.2 | NP_001036359.1 | 5 - 6 | 4289 | 87 | |
| NM_080364.4 | NP_525103.2 | 4 - 5 | 4551 | 87 | ||
More Information
Set Membership: |
|

bun
Genomic Map