Assay Details
| Gene Symbol: Mgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080378.4 | NP_525117.2 | 1 - 2 | 1027 | 77 | |
| GenBank mRNA | AB176552.1 | - | 1 - 2 | 466 | 77 | |
| BT032838.1 | - | 1 - 2 | 983 | 77 | ||
| BT124781.1 | - | 2 - 3 | 1858 | 77 | ||
More Information
Set Membership: |
|

Mgat1
Genomic Map