Assay Details
| Gene Symbol: Spn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259641.1 | NP_001246570.1 | 10 - 11 | 4977 | 87 | |
| NM_001259642.1 | NP_001246571.1 | 10 - 11 | 4977 | 87 | ||
| NM_001259643.1 | NP_001246572.1 | 10 - 11 | 4986 | 87 | ||
| NM_001259644.1 | NP_001246573.1 | 10 - 11 | 4977 | 87 | ||
| NM_001259645.1 | NP_001246574.1 | 10 - 11 | 4977 | 87 | ||
| NM_001259646.1 | NP_001246575.1 | 9 - 10 | 4956 | 87 | ||
| NM_001259647.1 | NP_001246576.1 | 9 - 10 | 4385 | 87 | ||
| NM_001259649.1 | NP_001246578.1 | 10 - 11 | 4977 | 87 | ||
| NM_001299992.1 | NP_001286921.1 | 10 - 11 | 4977 | 87 | ||
| GenBank mRNA | BT133165.1 | - | 10 - 11 | 4982 | 87 | |
More Information
Set Membership: |
|

Spn
Genomic Map