Assay Details
| Gene Symbol: Spn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259641.1 | NP_001246570.1 | 5 - 6 | 3315 | 83 | |
| NM_001259642.1 | NP_001246571.1 | 5 - 6 | 3315 | 83 | ||
| NM_001259643.1 | NP_001246572.1 | 5 - 6 | 3315 | 83 | ||
| NM_001259644.1 | NP_001246573.1 | 5 - 6 | 3315 | 83 | ||
| NM_001259645.1 | NP_001246574.1 | 5 - 6 | 3315 | 83 | ||
| NM_001259647.1 | NP_001246576.1 | 4 - 5 | 2723 | 83 | ||
| NM_001259648.1 | NP_001246577.1 | 5 - 6 | 3315 | 83 | ||
| NM_001259649.1 | NP_001246578.1 | 5 - 6 | 3315 | 83 | ||
| NM_001299992.1 | NP_001286921.1 | 5 - 6 | 3315 | 83 | ||
| NM_080516.3 | NP_536777.3 | 5 - 6 | 3315 | 83 | ||
| GenBank mRNA | BT133165.1 | - | 5 - 6 | 3311 | 83 | |
More Information
Set Membership: |
|

Spn
Genomic Map