Assay Details
| Gene Symbol: robo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272939.2 | NP_001259868.1 | 5 - 6 | 1276 | 55 | |
| NM_080531.4 | NP_536792.2 | 5 - 6 | 1276 | 55 | ||
| GenBank mRNA | BT150394.1 | - | 5 - 6 | 1272 | 55 | |
More Information
Set Membership: |
|

robo2
Genomic Map