Assay Details
| Gene Symbol: tty | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272837.2 | NP_001259766.1 | 7 - 8 | 2726 | 116 | |
| NM_001272839.2 | NP_001259768.1 | 6 - 7 | 2822 | 116 | ||
| NM_001272840.2 | NP_001259769.1 | 6 - 7 | 2882 | 116 | ||
| NM_080712.7 | NP_542443.4 | 7 - 8 | 2896 | 116 | ||
| NM_167744.4 | NP_728415.4 | 7 - 8 | 3494 | 116 | ||
| GenBank mRNA | AF184227.1 | - | 7 - 8 | 2813 | 116 | |
More Information
Set Membership: |
|

tty
Genomic Map