Assay Details
| Gene Symbol: Ing3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133088.1 | NP_573316.1 | 1 - 2 | 230 | 85 | |
| GenBank mRNA | BT088781.1 | - | 1 - 2 | 192 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ing3| Gene Symbol: Ing3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133088.1 | NP_573316.1 | 1 - 2 | 230 | 85 | |
| GenBank mRNA | BT088781.1 | - | 1 - 2 | 192 | 85 | |
Set Membership: |
|