Assay Details
| Gene Symbol: shop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133103.3 | NP_573331.1 | 1 - 2 | 1182 | 58 | |
| GenBank mRNA | AY069352.1 | - | 1 - 2 | 1167 | 58 | |
More Information
Set Membership: |
|

shop
Genomic Map