Assay Details
| Gene Symbol: htk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133102.3 | NP_573330.3 | 3 - 4 | 7411 | 78 | |
| NM_133104.3 | NP_573332.3 | 2 - 3 | 5316 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

htk| Gene Symbol: htk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133102.3 | NP_573330.3 | 3 - 4 | 7411 | 78 | |
| NM_133104.3 | NP_573332.3 | 2 - 3 | 5316 | 78 | ||
Set Membership: |
|