Assay Details
| Gene Symbol: CG43759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272768.1 | NP_001259697.1 | 6 - 7 | 3316 | 67 | |
| NM_001272769.1 | NP_001259698.1 | 6 - 7 | 3316 | 67 | ||
| NM_001298493.1 | NP_001285422.1 | 5 - 6 | 3129 | 67 | ||
| NM_133114.4 | NP_573342.1 | 6 - 7 | 3316 | 67 | ||
| GenBank mRNA | AY059450.1 | - | 6 - 7 | 3292 | 67 | |
More Information
Set Membership: |
|

CG43759
Genomic Map