Assay Details
| Gene Symbol: Ulp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133134.3 | NP_573362.1 | 2 - 3 | 2873 | 70 | |
| GenBank mRNA | BT030817.1 | - | 2 - 3 | 2782 | 70 | |
More Information
Set Membership: |
|

Ulp1
Genomic Map