Assay Details
| Gene Symbol: AIMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145193.2 | NP_660194.1 | 1 - 2 | 151 | 77 | |
| GenBank mRNA | AY113564.1 | - | 1 - 2 | 133 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AIMP3| Gene Symbol: AIMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145193.2 | NP_660194.1 | 1 - 2 | 151 | 77 | |
| GenBank mRNA | AY113564.1 | - | 1 - 2 | 133 | 77 | |
Set Membership: |
|