Assay Details
| Gene Symbol: crq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258894.1 | NP_001245823.1 | 2 - 3 | 283 | 62 | |
| NM_001272895.1 | NP_001259824.1 | 2 - 3 | 386 | 62 | ||
| NM_001272896.1 | NP_001259825.1 | 3 - 4 | 597 | 62 | ||
| NM_175943.2 | NP_787957.1 | 2 - 3 | 658 | 62 | ||
| NM_175944.3 | NP_787958.1 | 3 - 4 | 408 | 62 | ||
| GenBank mRNA | BT133259.1 | - | 2 - 3 | 659 | 62 | |
More Information
Set Membership: |
|

crq
Genomic Map