Assay Details
| Gene Symbol: tkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175976.2 | NP_787990.1 | 1 - 2 | 161 | 73 | |
| GenBank mRNA | BT133408.1 | - | 1 - 2 | 161 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tkv| Gene Symbol: tkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175976.2 | NP_787990.1 | 1 - 2 | 161 | 73 | |
| GenBank mRNA | BT133408.1 | - | 1 - 2 | 161 | 73 | |
Set Membership: |
|