Assay Details
| Gene Symbol: tkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175978.2 | NP_787992.1 | 1 - 2 | 1033 | 59 | |
| GenBank mRNA | AY060782.1 | - | 1 - 2 | 1012 | 59 | |
| L33784.1 | - | 1 - 2 | 432 | 59 | ||
| U11442.2 | - | 1 - 2 | 1051 | 59 | ||
More Information
Set Membership: |
|

tkv
Genomic Map