Assay Details
| Gene Symbol: und | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273370.2 | NP_001260299.1 | 2 - 3 | 1012 | 75 | |
| NM_001298854.1 | NP_001285783.1 | 2 - 3 | 1058 | 75 | ||
| NM_058025.5 | NP_477373.1 | 2 - 3 | 1178 | 75 | ||
| GenBank mRNA | AY047525.1 | - | 2 - 3 | 1055 | 75 | |
More Information
Set Membership: |
|

und
Genomic Map