Assay Details
| Gene Symbol: RpII33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058071.4 | NP_477419.1 | 1 | 351 | 67 | |
| GenBank mRNA | AJ133650.1 | - | 1 | 268 | 67 | |
| AY060644.1 | - | 1 | 312 | 67 | ||
More Information
Set Membership: |
|

RpII33
Genomic Map