Assay Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169462.2 | NP_001162933.1 | 4 - 5 | 1326 | 90 | |
| NM_058074.4 | NP_477422.2 | 5 - 6 | 1169 | 90 | ||
| NM_058075.4 | NP_477423.1 | 5 - 6 | 1057 | 90 | ||
| NM_058076.4 | NP_477424.1 | 3 - 4 | 827 | 90 | ||
| NM_134320.3 | NP_599147.1 | 5 - 6 | 1057 | 90 | ||
| NM_134321.3 | NP_599148.1 | 3 - 4 | 827 | 90 | ||
| NM_134322.3 | NP_599149.1 | 3 - 4 | 827 | 90 | ||
| GenBank mRNA | AF144232.1 | - | 4 - 5 | 600 | 90 | |
| AF161257.1 | - | 3 - 4 | 838 | 90 | ||
| AF161259.1 | - | 3 - 4 | 838 | 90 | ||
| BT025053.1 | - | 4 - 5 | 1255 | 90 | ||
More Information
Set Membership: |
|

Pten
Genomic Map