Assay Details
| Gene Symbol: esc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058083.4 | NP_477431.1 | 2 - 3 | 368 | 73 | |
| GenBank mRNA | AY069796.1 | - | 2 - 3 | 340 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

esc| Gene Symbol: esc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058083.4 | NP_477431.1 | 2 - 3 | 368 | 73 | |
| GenBank mRNA | AY069796.1 | - | 2 - 3 | 340 | 73 | |
Set Membership: |
|