Assay Details
Gene Symbol: gammaSnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058121.4 | NP_477469.1 | 1 - 2 | 135 | 68 | |
GenBank mRNA | AF177988.1 | - | 1 - 2 | 65 | 68 | |
AY119512.1 | - | 1 - 2 | 85 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
