Assay Details
| Gene Symbol: gammaSnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058121.4 | NP_477469.1 | 2 - 3 | 203 | 85 | |
| GenBank mRNA | AF177988.1 | - | 2 - 3 | 133 | 85 | |
| AY119512.1 | - | 2 - 3 | 153 | 85 | ||
More Information
Set Membership: |
|

gammaSnap1
Genomic Map