Assay Details
| Gene Symbol: fs(1)K10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058143.4 | NP_477491.1 | 1 - 2 | 1468 | 73 | |
| GenBank mRNA | AY060415.1 | - | 1 - 2 | 1449 | 73 | |
More Information
Set Membership: |
|

fs(1)K10
Genomic Map