Assay Details
| Gene Symbol: png | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058144.3 | NP_477492.1 | 1 | 1056 | 148 | |
| GenBank mRNA | AF306498.1 | - | 1 | 1068 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

png| Gene Symbol: png | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058144.3 | NP_477492.1 | 1 | 1056 | 148 | |
| GenBank mRNA | AF306498.1 | - | 1 | 1068 | 148 | |
Set Membership: |
|