Assay Details
| Gene Symbol: Fs(2)Ket | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259179.2 | NP_001246108.1 | 2 - 3 | 357 | 76 | |
| NM_001259180.2 | NP_001246109.1 | 3 - 4 | 539 | 76 | ||
| NM_001259181.2 | NP_001246110.1 | 2 - 3 | 357 | 76 | ||
| NM_001259182.2 | NP_001246111.1 | 2 - 3 | 357 | 76 | ||
| NM_058148.5 | NP_477496.1 | 2 - 3 | 357 | 76 | ||
| GenBank mRNA | AF222745.1 | - | 2 - 3 | 348 | 76 | |
| AY118657.1 | - | 2 - 3 | 349 | 76 | ||
More Information
Set Membership: |
|

Fs(2)Ket
Genomic Map