Assay Details
| Gene Symbol: Fs(2)Ket | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259179.2 | NP_001246108.1 | 5 - 6 | 2558 | 68 | |
| NM_001259180.2 | NP_001246109.1 | 6 - 7 | 2740 | 68 | ||
| NM_001259181.2 | NP_001246110.1 | 5 - 6 | 2558 | 68 | ||
| NM_001259182.2 | NP_001246111.1 | 5 - 6 | 2558 | 68 | ||
| NM_058148.5 | NP_477496.1 | 5 - 6 | 2558 | 68 | ||
| GenBank mRNA | AF222745.1 | - | 5 - 6 | 2549 | 68 | |
| AY118657.1 | - | 5 - 6 | 2550 | 68 | ||
More Information
Set Membership: |
|

Fs(2)Ket
Genomic Map