Assay Details
| Gene Symbol: ft | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058149.4 | NP_477497.1 | 6 - 7 | 7135 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ft| Gene Symbol: ft | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058149.4 | NP_477497.1 | 6 - 7 | 7135 | 104 | |
Set Membership: |
|