Assay Details
| Gene Symbol: fzy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058153.4 | NP_477501.1 | 1 - 2 | 462 | 74 | |
| GenBank mRNA | BT029432.1 | - | 6330 | 74 | ||
| BT056243.1 | - | 1 - 2 | 462 | 74 | ||
| U22419.1 | - | 1 - 2 | 427 | 74 | ||
More Information
Set Membership: |
|

fzy
Genomic Map