Assay Details
| Gene Symbol: CG8051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298505.1 | NP_001285434.1 | 1 - 2 | 459 | 105 | |
| NM_133149.3 | NP_573377.1 | 2 - 3 | 740 | 105 | ||
| GenBank mRNA | AY069861.1 | - | 2 - 3 | 708 | 105 | |
More Information
Set Membership: |
|

CG8051
Genomic Map