Assay Details
| Gene Symbol: CG8051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298505.1 | NP_001285434.1 | 3 - 4 | 1885 | 66 | |
| NM_133149.3 | NP_573377.1 | 4 - 5 | 2166 | 66 | ||
More Information
Set Membership: |
|

CG8051
Genomic Map