Assay Details
| Gene Symbol: Ance-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133164.4 | NP_573392.2 | 3 - 4 | 1987 | 72 | |
| GenBank mRNA | BT044428.1 | - | 3 - 4 | 1946 | 72 | |
More Information
Set Membership: |
|

Ance-5
Genomic Map