Assay Details
| Gene Symbol: Ubqn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298528.1 | NP_001285457.1 | 1 - 2 | 289 | 68 | |
| NM_134500.4 | NP_608344.1 | 1 - 2 | 289 | 68 | ||
| GenBank mRNA | AY128447.1 | - | 1 - 2 | 258 | 68 | |
More Information
Set Membership: |
|

Ubqn
Genomic Map