Assay Details
| Gene Symbol: pico | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134519.3 | NP_608363.1 | 1 - 2 | 1199 | 58 | |
| GenBank mRNA | BT124745.1 | - | 1 - 2 | 516 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pico| Gene Symbol: pico | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134519.3 | NP_608363.1 | 1 - 2 | 1199 | 58 | |
| GenBank mRNA | BT124745.1 | - | 1 - 2 | 516 | 58 | |
Set Membership: |
|