Assay Details
| Gene Symbol: RhoGAP19D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169338.2 | NP_001162809.1 | 14 - 15 | 5659 | 77 | |
| NM_001258851.1 | NP_001245780.1 | 15 - 16 | 5737 | 77 | ||
| NM_001298567.1 | NP_001285496.1 | 14 - 15 | 5950 | 77 | ||
| NM_001298568.1 | NP_001285497.1 | 13 - 14 | 5252 | 77 | ||
| NM_134552.6 | NP_608396.2 | 13 - 14 | 5875 | 77 | ||
More Information
Set Membership: |
|

RhoGAP19D
Genomic Map