Assay Details
| Gene Symbol: RhoGAP19D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169338.2 | NP_001162809.1 | 4 - 5 | 1444 | 53 | |
| NM_001258851.1 | NP_001245780.1 | 4 - 5 | 1444 | 53 | ||
| NM_001298567.1 | NP_001285496.1 | 4 - 5 | 1444 | 53 | ||
| NM_001298568.1 | NP_001285497.1 | 2 - 3 | 959 | 53 | ||
| NM_134552.6 | NP_608396.2 | 4 - 5 | 1444 | 53 | ||
More Information
Set Membership: |
|

RhoGAP19D
Genomic Map