Assay Details
| Gene Symbol: RhoGAP19D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169338.2 | NP_001162809.1 | 10 - 11 | 3643 | 111 | |
| NM_001258851.1 | NP_001245780.1 | 11 - 12 | 3721 | 111 | ||
| NM_001298567.1 | NP_001285496.1 | 11 - 12 | 3721 | 111 | ||
| NM_001298568.1 | NP_001285497.1 | 9 - 10 | 3236 | 111 | ||
| NM_134552.6 | NP_608396.2 | 10 - 11 | 3646 | 111 | ||
More Information
Set Membership: |
|

RhoGAP19D
Genomic Map