Assay Details
| Gene Symbol: shakB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078701.3 | NP_523425.2 | 1 | 151 | 99 | |
| NM_134566.3 | NP_608410.2 | 1 | 151 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_078701.3 transcripts.
Set Membership: |
|

shakB
Genomic Map