Assay Details
| Gene Symbol: Cda5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103574.3 | NP_001097044.2 | 3 - 4 | 286 | 64 | |
| NM_001103575.2 | NP_001097045.1 | 2 - 3 | 178 | 64 | ||
| NM_001258876.2 | NP_001245805.1 | 2 - 3 | 415 | 64 | ||
| NM_001258877.2 | NP_001245806.1 | 2 - 3 | 164 | 64 | ||
| NM_001258878.2 | NP_001245807.1 | 2 - 3 | 195 | 64 | ||
| NM_001258879.2 | NP_001245808.1 | 3 - 4 | 667 | 64 | ||
| NM_164353.2 | NP_722589.1 | 2 - 3 | 184 | 64 | ||
| NM_164354.3 | NP_722590.2 | 2 - 3 | 184 | 64 | ||
| GenBank mRNA | BT057995.1 | - | 2 - 3 | 159 | 64 | |
More Information
Set Membership: |
|

Cda5
Genomic Map