Assay Details
| Gene Symbol: Cda5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103574.3 | NP_001097044.2 | 8 - 9 | 4131 | 81 | |
| NM_001103575.2 | NP_001097045.1 | 6 - 7 | 1470 | 81 | ||
| NM_001258877.2 | NP_001245806.1 | 7 - 8 | 3709 | 81 | ||
| NM_164353.2 | NP_722589.1 | 6 - 7 | 1476 | 81 | ||
| NM_164354.3 | NP_722590.2 | 6 - 7 | 1476 | 81 | ||
| GenBank mRNA | BT057995.1 | - | 6 - 7 | 1451 | 81 | |
More Information
Set Membership: |
|

Cda5
Genomic Map