Assay Details
| Gene Symbol: Cda5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103574.3 | NP_001097044.2 | 6 - 7 | 3591 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cda5| Gene Symbol: Cda5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103574.3 | NP_001097044.2 | 6 - 7 | 3591 | 65 | |
Set Membership: |
|