Assay Details
| Gene Symbol: Cda5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164354.3 | NP_722590.2 | 7 - 8 | 1916 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cda5| Gene Symbol: Cda5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164354.3 | NP_722590.2 | 7 - 8 | 1916 | 112 | |
Set Membership: |
|