Assay Details
| Gene Symbol: cbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164384.2 | NP_722636.1 | 1 - 2 | 282 | 98 | |
| GenBank mRNA | BT099918.1 | - | 1 - 2 | 276 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cbt| Gene Symbol: cbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164384.2 | NP_722636.1 | 1 - 2 | 282 | 98 | |
| GenBank mRNA | BT099918.1 | - | 1 - 2 | 276 | 98 | |
Set Membership: |
|