Assay Details
| Gene Symbol: Nnf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164388.3 | NP_722652.1 | 2 - 3 | 393 | 97 | |
| GenBank mRNA | BT015998.1 | - | 2 - 3 | 384 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nnf1b| Gene Symbol: Nnf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164388.3 | NP_722652.1 | 2 - 3 | 393 | 97 | |
| GenBank mRNA | BT015998.1 | - | 2 - 3 | 384 | 97 | |
Set Membership: |
|