Assay Details
| Gene Symbol: Obp22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014457.3 | NP_001014457.1 | 1 - 2 | 71 | 67 | |
| NM_164447.3 | NP_722746.2 | 1 - 2 | 71 | 67 | ||
| GenBank mRNA | AF457144.1 | - | 1 - 2 | 63 | 67 | |
More Information
Set Membership: |
|

Obp22a
Genomic Map