Assay Details
| Gene Symbol: Kebab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272971.1 | NP_001259900.1 | 3 - 4 | 614 | 69 | |
| NM_164451.2 | NP_722756.1 | 3 - 4 | 611 | 69 | ||
| GenBank mRNA | AY051677.1 | - | 3 - 4 | 580 | 69 | |
| BT126028.1 | - | 3 - 4 | 563 | 69 | ||
More Information
Set Membership: |
|

Kebab
Genomic Map