Assay Details
| Gene Symbol: tho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272976.1 | NP_001259905.1 | 3 - 4 | 801 | 94 | |
| NM_134802.4 | NP_608646.3 | 3 - 4 | 801 | 94 | ||
| NM_164454.2 | NP_722763.1 | 3 - 4 | 801 | 94 | ||
| GenBank mRNA | AJ556820.2 | - | 3 - 4 | 645 | 94 | |
| BT031136.1 | - | 3 - 4 | 789 | 94 | ||
More Information
Set Membership: |
|

tho2
Genomic Map